Graduated pipettes, SILBERBRAND, with piston, total delivery
Supplier: Brand
612-1001EA 17.7 EUR
BRND31106
Graduated pipettes, SILBERBRAND, with piston, total delivery
AR-Glas®.
  • Calibrated to deliver 'Ex'
  • Marks and inscriptions in high contrast white enamel
  • Integrated piston pipetting aid with retaining spring
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.