You Searched For: Pipette Washers
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Brand
Description: This system enables trouble-free, gentle cleaning of pipettes with repetitive siphon rinsing.
Supplier: Thermo Fisher Scientific
Description: These conveniently packaged sets contain a basket, a jar and a washer. A larger size is available to accomodate up to 32" (81 cm) long pipettes.
Supplier: Thermo Fisher Scientific
Description: For washing and rinsing of pipettes. Insert a filled pipette basket. Attach it to your sink water or DI tap and drain for repeated, automatic fill and...
Supplier: KARTELL
Description: Rinsing sets for cleaning pipettes up to 600 mm length and Mohr burettes gently and efficiently. The entire washing system consists of a washer, a pip...
Catalog Number: (BURK9650-0650)
Supplier: BÃœRKLE
Description: For easy and gentle cleaning and safe storage of pipettes. A complete system suitable for every lab.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom