VWR® Powerpette Plus Pipette Controller
Supplier: Avantor
Powerpette
JENC266-000EA 318 EUR
JENC266-000
VWR® Powerpette Plus Pipette Controller
Pipettes Pipette Controllers
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
  • Speed of suction and dispensing controlled through the finger triggers
  • Fully-charged has approximately 8 hours of continuous use available
  • Low battery light illuminates when one hour's power remains
Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Three aspirate and dispense modes. High and low settings both have variable speed. Gravity dispense mode is designed for use with ‘To Deliver' (TD) pipettes.
Ordering information: Supplied with 0,45 µm and 0,2 µm hydrophobic filters, as standard, for effective protection against airborne contamination of the unit during aspiration, and of the samples during dispensing; 2×1,2 V nickel metal hydride batteries and UK-charger.