You Searched For: Mops
145  results were found
SearchResultCount:"145"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Foam with 100% polyester cover. For applying disinfectants during pharmaceutical production or for aseptic processes. The flat mop head is ideally sui...
Supplier: VWR Collection
Description: This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for...
Supplier: VWR Collection
Description: Mops suitable for wet wiping in sterile areas and cleanrooms. One-piece, made from highly absorbent polyurethane. These coated mop heads are ideal for...
Supplier: VWR Collection
Description: 100% polyester string mops. Double clear poly bagged for cleanroom use. Good chemical sorbancy. Ideal for application of cleaning solutions on fl...
Clearance
Supplier: VWR Collection
Description: Mop heads are made of polyester fabric laminated to clean ester foam and are free of any cellulosic contaminants. Developed for cleaning ceilings and ...
Sale
Supplier: CONTEC
Description: Designed to simplify the cleanroom mopping process, Contec's Hands free mop head saturation system allows mop heads to be presaturated prior to being ...
Sale
Catalog Number: (141-1392)
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Mop for all smooth hard floorings. Made up of 100% PES super microfiber.
UOM: 1 * 1 items
Supplier: CONTEC
Description: These lightweight, polyester disposable mops are excellent for applying disinfectants and for wet cleaning of vertical surfaces, ceilings, and walls i...
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Mops for all smooth and textured hard floorings and safety tiles. Made up of 100% PES super microfiber with soft-brush.
Supplier: CONTEC
Description: EasyReachâ„¢ is recommended for effective cleaning and disinfection of surfaces of separative devices such as: Isolators, restricted access barrier syst...
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Mops made from 100% polyester microfibre yarn (0,5 De)
Supplier: CONTEC
Description: Contec’s EasyCurve™ Mop was designed specifically for critical and controlled environments, now available with a ribbed microfiber face giving users s...
Supplier: CONTEC
Description: The DualCleanâ„¢ Softwall Strip Cleaner is ideal for cleaning strip curtains and barriers in all environments from clean workspace enclosure to ISO Clas...
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Autoclavable systembox EasyMop GMP and MopJump-System:
The newly developed 'EasyMop GMP' system box ensures compliance with regulations g...
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Squeegee mops and handles for hygienically sensitive areas.
Supplier: CONTEC
Description: QuickTaskâ„¢ mopping system. The QuickTaskâ„¢ product line consists of a variety of knitted polyester, polyester/cellulose, microfibre/foam and microfibre...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 145
no targeter for Bottom