VWR®, Cover Glasses, Square
Supplier: VWR
VWR®
631-0121EA 29 EUR
631-0121 631-0120 631-0127 631-0126 631-0125 631-0124
VWR®, Cover Glasses, Square
Cover Glasses
Made of pure white Hydrolytic Class 1 glass.
  • No bubbles or flaws
  • Cleaned and polished
Thickness: 0,08 - 0,19 mm
Packaging: In a plastic box with hinged lid.