Thermo-Tube PCR plates, Thermo-Fast®
Supplier: Thermo Scientific
732-4857EA 246 EUR
ADVAAB-0731
Thermo-Tube PCR plates, Thermo-Fast®
PP. 96-well plate suitable for use with most thermal cyclers, including PerkinElmer 9600 and 9700.
  • 12 rows of 8 Thermo-Strips, pre-assembled into a strip retainer plate
  • Block stops on the underside of the retainer to allow easy removal from the thermal cycler
  • Maximum well volume 0,2 ml
  • Certified free from DNase, RNase and human genomic DNA