You Searched For: PCR Plates
87  results were found
SearchResultCount:"87"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: PP. These PCR plates are compatible with most thermal cyclers.
Sale
Supplier: VWR Collection
Description: Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results.
Sale
Catalog Number: (732-3492)
Supplier: VWR Collection
Description: UltraFlux® PCR Plates are available in variety of styles to fit most thermal cyclers in the market.
UOM: 1 * 10 items
Supplier: VWR Collection
Description: VWR® 96- and 384-well Real-Time PCR plates are compatible with most leading Real-Time PCR instruments including most BioRad, Applied Biosystems, Roche...
Low Manufacturing Impact
Sustainable Packaging Sale
Supplier: Avantor
Description: J.T.Baker® PCR plates are constructed with two plastic components. A polycarbonate ultra-rigid frame for automation and polypropylene ultra-thin wells...
Sale
Catalog Number: (732-3231)
Supplier: VWR Peqlab
Description: PP. Semi-skirted plate especially designed for Real-Time PCR applications.
UOM: 1 * 25 items
Supplier: VWR Collection
Description: VWR® 96-Well PCR Plates are compatible with most leading PCR and real-time PCR instruments.
Low Manufacturing Impact
Sustainable Packaging Sale
Supplier: VWR Peqlab
Description: PP, non-skirted. Low profile plates significantly reduce the dead space between the film and the sample, which reduces the risk of sample condensation...
Supplier: VWR Peqlab
Description: PP, autoclavable. Low profile, semi-skirted plates for use on the ABI Fast Block system or the Roche® LightCycler® 480, offering optimal instrument fi...
Supplier: VWR Peqlab
Description: PP. Compatible with all major thermal cyclers.
Catalog Number: (732-3237)
Supplier: VWR Peqlab
Description: PP, autoclavable. These high quality, 384-well PCR plates are suitable for high throughput applications, and are designed to be compatible with roboti...
UOM: 1 * 50 items
Supplier: VWR Peqlab
Description: PP, non-skirted. Standard 96-well PCR plates, suitable for a wide range of 0,2 ml thermal cycler blocks.
Catalog Number: (732-3225)
Supplier: VWR Peqlab
Description: PP. Low profile plates significantly reduce the dead space between the film and the sample, which reduces the risk of sample condensation for enhanced...
UOM: 1 * 25 items
Catalog Number: (732-3217)
Supplier: VWR Peqlab
Description: PP. Semi-skirted plate with 24 wells for medium density sample throughput.
UOM: 1 * 50 items
Supplier: Thermo Fisher Scientific
Description: PP. The PCR detection plate features a flat deck for improved handling and sealing.
Sale
Catalog Number: (732-4912)
Supplier: Thermo Fisher Scientific
Description: Moulded from ultra low retention PP that assists in preventing adhesion of samples to the side walls. Due to the increased well volume, post-PCR appli...
UOM: 1 * 100 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 87
no targeter for Bottom