Microarray plates, 384-well
Supplier: Thermo Scientific
ABgene®
732-4941EA 236 EUR
MATRAB-1055 732-0086
Microarray plates, 384-well
PP. These 384 well plates are ideal for use as source plates for arraying and other low volume liquid handling applications.
  • 384 round wells with conical well bottom shape
  • Raised well rims for effective sealing and reduced contamination risk
  • Precision moulded with flat profile and strengthening ribs
  • Alphanumeric grid referencing and cut-away corner
  • Fully skirted and within SBS footprint recommendations for compatibility with automated systems
Well volume: 70 µl