You Searched For: Insert Plates and Inserts
32  results were found
SearchResultCount:"32"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: PC, sterile, with lid. These insert plates feature a thin, semi-transparent polycarbonate membrane with pore sizes from 0,1 to 12,0 μm.
Supplier: VWR Collection
Description: Polyester, sterile, with lid. These insert plates feature a thin, microscopically transparent polyester membrane that is tissue culture-treated for op...
Catalog Number: (734-0154)
Supplier: Corning
Description: Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such ...
UOM: 1 * 24 items
Supplier: Corning
Description: The Corning® BioCoat® Angiogenesis systems facilitate investigation of compound effects on endothelial cell invasion, migration, and tubulogenesis. Th...
Supplier: Corning
Description: Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such ...
Supplier: Corning
Description: Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.
Supplier: Corning
Description: The Corning® FluoroBlok™ HTS 24-multiwell insert system contains an automation-friendly, 24-multiwell cell culture membrane insert suitable for both m...
Supplier: Corning
Description: PS, tissue culture-treated, sterile, non-pyrogenic, with lid. Specifically designed for use with Falcon® or Corning® BioCoat™ cell culture inserts, so...
Supplier: Corning
Description: The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based...
Supplier: Corning
Description: Corning® FluoroBlok™ HTS 96-multiwell insert system is a cell culture assay platform designed for automation. The one-piece insert housing and fluores...
Catalog Number: (734-1053)
Supplier: Corning
Description: Corning® BioCoat™ FluoroBlok™ fibronectin-coated cell culture inserts packaged ready to use in 24-well Falcon® companion plates.
UOM: 1 * 24 items
Supplier: Corning
Description: The Falcon® HTS 24-multiwell insert system is designed to automate many commonly used membrane-based cell assays and increase the productivity and thr...
Catalog Number: (734-1079)
Supplier: Corning
Description: Multiwell insert system with feeder tray and lid, fibrillar collagen, Corning® BioCoat®, Non sterile, Plate format: 24-well insert plates, Pore size (µm): 1,00 µm
UOM: 1 * 1 items
Supplier: Corning
Description: Permit real-time analysis of cell culture compound permeability (1 µm), cell migration (3 µm or 8 µm), and invasion (8 µm).
Supplier: Corning
Description: 75 mm Transwell® with 0,4 µm pore polycarbonate membrane insert, sterile.
Supplier: Corning
Description: Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 32
no targeter for Bottom