You Searched For: Microplates
538  results were found
SearchResultCount:"538"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (732-3231)
Supplier: VWR Peqlab
Description: PP. Semi-skirted plate especially designed for Real-Time PCR applications.
UOM: 1 * 25 items
Supplier: Avantor
Description: Made of virgin PP to minimise extractables.
Sale
Supplier: VWR Collection
Description: VWR® multiwell cell culture plates are available with a choice of surface treatments.
Supplier: VWR Collection
Description: Designed specifically for immunoassay applications.
Sale
Supplier: VWR Collection
Description: Manufactured from high quality crystal polystyrene. They are ideal for routine assays, mixing and storage.
Sale
Supplier: VWR Collection
Description: Polyester, sterile, with lid. These insert plates feature a thin, microscopically transparent polyester membrane that is tissue culture-treated for op...
Sale
Supplier: VWR Collection
Description: PP. These PCR plates are compatible with most thermal cyclers.
Catalog Number: (732-0612)
Supplier: VWR Collection
Description: PP. These plates hold a full 2,5 ml (2,2 ml with sealing mat in place) and work as storage plates or reservoirs.
UOM: 1 * 10 items
Low Manufacturing Impact
Sustainable Packaging
Supplier: VWR Collection
Description: Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results.
Sale
Supplier: VWR Collection
Description: PC, sterile, with lid. These insert plates feature a thin, semi-transparent polycarbonate membrane with pore sizes from 0,1 to 12,0 μm.
Supplier: VWR Collection
Description: Single-well cell culture plates, made from clear PS, are available plasma-treated for consistent cell attachment and growth, or non-treated for use in...
Supplier: VWR Collection
Description: VWR® 96- and 384-well Real-Time PCR plates are compatible with most leading Real-Time PCR instruments including most BioRad, Applied Biosystems, Roche...
Low Manufacturing Impact
Sustainable Packaging Sale
Supplier: Avantor
Description: Solid Phase Extraction (SPE) is commonly used as a highly selective sample clean-up or sample enrichment method prior to subsequent analyses. J.T.Bake...
Supplier: VWR Collection
Description: These deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal...
Supplier: Avantor
Description: These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as ...
Sale
Supplier: Avantor
Description: Available in PP for automated screening and storage systems.
Sale
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 538
no targeter for Bottom