Reusable half masks against gases, vapours and particulates, 4000 series
Supplier: 3M
111-0041EA 34.6 EUR
111-0041 MMMA4255CS 111-0044
Reusable half masks against gases, vapours and particulates, 4000 series
Respirators Disposable Respirators Gas or Gas and Particulate Respirators
The resusable half mask with integrated filters provides effective protection combined with exceptional comfort.
  • Fully disposable
  • Integral construction offers reliable protection
  • Parabolic valve reduces breathing resistance whilst preventing heat build-up
Weight: 222 g
4277
Certifications: EN 405:2001+A1:2009
  • EN 405: 2002 FFABE1P3D
  • Protects against fine dusts, oil and water-based mists
  • Additional gas and vapour relief: organic gases, inorganic gases, acid gases and organic vapours
  • Maximum use level: Assigned protection factor (APF): 20× WEL for particles, 10× WEL or 1000 ppm, whichever is lower for organic gases, inorganic gases, acid gases and organic vapours
  • EN 405: 2002 FFABEKP3D
  • Protects against fine dusts, oil and water-based mists
  • Additional gas and vapour relief: organic gases, inorganic gases, acid gases, ammonia and organic vapours
  • Maximum use level: Assigned protection factor (APF): 20× WEL for particles, 10× WEL or 1000 ppm, whichever is lower for organic gases, inorganic gases, acid gases, ammonia and organic vapours
Caution: *One size fits all