Disposable particle filtering respirators, FFP1/FFP2, Classical and Comfort series
Supplier: 3M
Comfort Classical
111-3001EA 49 EUR
111-3001 111-0100 111-0102 111-0101
Disposable particle filtering respirators, FFP1/FFP2, Classical and Comfort series
Respirators Disposable Respirators Particulate Respirators
The convex shape of the face mask with twin strap design, foam nose seal and aluminium nose clip ensures a good face seal over a wide range of face sizes.
  • Provides lightweight, effective, comfortable and hygienic respiratory protection
  • Durable, collapse resistant inner shell
  • Exempt from costly time-consuming maintenance requirements
8812 (valved)
Certifications: EN 149:2001; 89/686; 10/11B
  • Minimum of 78% protection against dust, water-based and non volatile liquid aerosols down to 0,5 micron
  • The exhalation valve reduces heat build-up to offer comfortable protection particularly in hot and humid conditions
  • Maximum use level: Assigned Protection Factor: 4× WEL
  • Minimum of 92% protection against dust, water-based and non volatile liquid aerosols down to 0,5 micron
  • The exhalation valve reduces heat build-up to offer comfortable protection particularly in hot and humid conditions
  • Maximum use level: Assigned Protection Factor: 10× WEL
Caution: * One size fits all