You Searched For: Masks
49  results were found
SearchResultCount:"49"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (76470-110)
Supplier: VWR Collection
Description: VWR® made in USA sterile masks are an ideal option for single-use needs.
UOM: 1 * 200 items
Supplier: VWR Collection
Description: Face masks made from 3 ply, non woven PP featuring a nose bridge and ear loops or ties.
Sale
Supplier: VWR Collection
Description: Cleanroom, duck bill face masks feature superior particle and bacterial filtration efficiency plus low particle shedding. These moisture resistant, cl...
Sale
Supplier: VWR Collection
Description: Maximum cleanroom masks feature three ply construction for superior particle and bacterial filtration efficiency. These masks are latex-free.
Sale
Catalog Number: (114-9101)
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: Hygiene masks made from 3-ply construction of PP and Meltblown PP.
UOM: 1 * 50 items
Supplier: VWR Collection
Description: Cleanroom masks made from non woven fabric featuring ties.
Sale
Supplier: VWR Collection
Description: Cleanroom face masks are manufactured with non woven polypropylene/polyethylene (ES) / melt-blown polypropylene (MBPP) materials.
Sale
Catalog Number: (113-0713)
Supplier: VWR Collection
Description: This face mask, with nose bridge, is made from two ply, non woven PP.
UOM: 1 * 50 items
Catalog Number: (HOIT115688)
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: Face mask with comfortable elastic ear loops for easy donning.
UOM: 1 * 10.000 items
Supplier: DESCO
Description: These facemasks constructed of a lightweight dissipative material which is made from texturised polyester and a minimum of 9% carbon nylon monofilimen...
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: Sterile cleanroom facemasks.
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: These masks are with 'no fog' adhesive strip.
Sale
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: Sterile cleanroom face masks feature Tyvek® side extensions to prevent skin exposure. Double elastic headband fastening makes it easier to put on comp...
Catalog Number: (113-8158)
Supplier: Nitritex
Description: Non sterile soft face veil with headband and nosepiece manufactured using ultrasonic bonding.
UOM: 1 * 1.000 items
Catalog Number: (113-9994)
Supplier: 3M MEDICAL
Description: These flat pleated surgical masks with adjustable tie provide comfort, breathability and maximum protection.
UOM: 1 * 100 items
Supplier: Nitritex
Description: The sterile pouch style mask has knitted headbands and all seams are ultrasonically bonded. The mask has inner and outer facing layers constructed of ...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 49
no targeter for Bottom