You Searched For: Laboratory Jacks
11  results were found
SearchResultCount:"11"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: High quality, stable and inexpensive range of lifting platforms for a variety of uses. With plates made from matt, brushed, non rusting stainless stee...
Sale
Supplier: VWR Collection
Description: Anodised aluminium. With M10 threaded hole in the middle of the top plate for attaching retort rods (starting from item number 245-0006).
Supplier: GRAUER RUDOLF
Description: These laboratory jacks are made of anodised aluminium for corrosion resistance and resistance to many chemicals.
Supplier: USBECK Laborgeräte
Description: Stainless steel plates, slip-proof rubber feet. With 10 mm hole in the top plate for attaching stand rods.
Supplier: BOCHEM
Description: These lab jacks feature an improved scissor technology with reduced tolerances. The scissor construction is made of ¹⁸/₁₀ stainless steel.
Supplier: BOCHEM
Description: Aluminium or anodised aluminium. These lab jacks feature an improved scissor technology with reduced tolerances. The scissor construction is made of 1...
Catalog Number: (245-0025)
Supplier: BOCHEM
Description: Stainless steel ¹⁸/₁₀. Remote control operated lab jack with integrated motor for use in hoods.
UOM: 1 * 1 items
Catalog Number: (HECH42011010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: This lab jack is made of powder-coated aluminioum alloy.
UOM: 1 * 1 items
Catalog Number: (BELAF183910001)
Supplier: Bel-Art Products, a Part of SP
Description: This corrosion resistat lab jack is molded of strong PC and assembled with no metal parts. It supports gradient makers, hot plates, flasks, beakers, b...
UOM: 1 * 1 items
Supplier: JUCHHEIM Laborgerate GmbH
Description: These hydraulic lifting stages feature a robust construction in light alloy or stainless steel.
Supplier: JUCHHEIM Laborgerate GmbH
Description: Aluminium alloy, with epoxy resin coating. These manually operated lifting stages provide high stablity.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 11 of 11
no targeter for Bottom