Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "adhesive-labels"
https://ru.vwr.com/stibo/smallweb/std.lang.all/48/81/18944881.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"35"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR labels are ideal for all cryostorage and can also be us...
Supplier:  VWR Collection
Description:   The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage and can also be u...
Supplier:  BRADY & CO LTD, W. H.
Description:   Write-On Labels provide easy identification for calibration, testing, and repair of tools and equipment.
Supplier:  WILMAD/LABGLASS/LUREX MFG
Description:   These labels are a handy alternative to marking NMR or EPR sample tubes. Each fits the circumference of the tube precisely with no overlapping to guar...
UOM:  1 * 480 items
Supplier:  BÜRKLE
Description:   Designed to seal the holes in sacks or boxes created when samples are taken, the strong adhesive on the ‘close-it®’ label allows adhesion to surfaces ...
Supplier:  BÜRKLE
Description:   Standard close-it® and close-it® food & pharma control seals are also available on a continuous roll. This offers the flexibility to provide a small l...
Supplier:  Sampling Systems
Description:   Labels, Quality Control, Dimensions: 150×150 mm, Yellow
Supplier:  BRADY & CO LTD, W. H.
Description:   Made of self destructive polyolefin, in German language. These inspection date labels can be used wherever regular checks on equipment and machinery a...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 35
  1  2  3  4  5  Next