You Searched For: Barrels, Kegs, Drums
32  results were found
SearchResultCount:"32"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (ENTE4B0100-0006)
Supplier: ENTEGRIS INC
Description: HDPE, triple layer.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: HDPE, translucent.
Supplier: BÃœRKLE
Description: HDPE white containers with red screw lid.
Supplier: Sampling Systems
Description: These stainless steel containers are made from high-quality 316L stainless steel and are ideal for almost all aplications, including GMP use.
Supplier: Kautex
Description: HDPE. With lid and clamping ring.
Supplier: BÃœRKLE
Description: Tubs, round, made of HDPE. For every purpose, with strong edge and practical carrying handles for safe transport. A lid can be delivered for every tub...
Supplier: Sampling Systems
Description: These drums which incorporate a sloping base allow the container to drain completely.
Supplier: Sampling Systems
Description: These stainless steel containers from 2 litres up to 300 litres are designed for storing and processing powders and liquids.
Supplier: Sampling Systems
Description: These drums are used for the storage and processing of powders and liquids.
Supplier: NEW PIG
Description: Tough enough for rough handling, guaranteed. Ship your haz waste in our UN rated overpacks to stay compliant with regs.
Supplier: Sampling Systems
Description: Made of stainless steel.
Supplier: CURTEC
Description: HDPE drums, with screw lid.
Supplier: LAMAPLAST
Description: Drums, made of HDPE, blue, UN approved, with lid.
Supplier: LAMAPLAST
Description: Drums, made of HDPE, white.
Supplier: CURTEC
Description: Drums, HDPE, nestable, white with blue screw lid.
Supplier: CURTEC
Description: Drums, made of UV safe HDPE, with screw lid. For storage and shipment of UV sensitive powders, pastes and liquids.
Sale
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 32
no targeter for Bottom