You Searched For: Skincare
124  results were found
SearchResultCount:"124"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GOJO Industries - Europe Ltd
Description: PURELL® VF+ Hygienic Hand Rub offers unprecedented sanitising while preserving healthy skin. Product benefits include a short contact time and an exce...
Sale
Supplier: GOJO Industries - Europe Ltd
Description: Exceptionally mild, unfragranced foam hand soap.
Safer Human and Environmental Health
Supplier: GOJO Industries - Europe Ltd
Description: Unfragranced and colourless foam hand soap.
Safer Human and Environmental Health
Supplier: GOJO Industries - Europe Ltd
Description: Remarkably mild soap formulation removes more than 99% of dirt and germs.
Safer Human and Environmental Health
Water Efficient
Supplier: GOJO Industries - Europe Ltd
Description: A hygienic, no-rinse hand rub that feels great to use and is clinically proven to maintain skin health.
Sustainable Materials
Supplier: GOJO Industries - Europe Ltd
Description: The quat-free GOJO® antimicrobial plus foam hand wash is gentle on skin and formulated to be highly effective.
Sale
Catalog Number: (GOJO5495-02-EEU)
Supplier: GOJO Industries - Europe Ltd
Description: PURELL® VF+ hygienic hand rub offers unprecedented sanitising whilst preserving healthy skin. Product benefits include a short contact time and an exc...
UOM: 1 * 2 items
MSDS
Supplier: GOJO Industries - Europe Ltd
Description: Unfragranced and colourless foam hand soap.
Catalog Number: (GOJO8848-03-EEU)
Supplier: GOJO Industries - Europe Ltd
Description: The quat-free GOJO® Antimicrobial Plus Foam Handwash is gentle on skin and formulated to be highly effective.
UOM: 1 * 3 items
Blocked
Supplier: GOJO Industries - Europe Ltd
Description: A no-rinse hygienic hand rub that feels great to use and is clinically proven to maintain skin health.
Supplier: GOJO Industries - Europe Ltd
Description: Remarkably mild soap formulation removes more than 99% of dirt and germs.
Safer Human and Environmental Health
Water Efficient
Catalog Number: (GOJO7335-04-EEU)
Supplier: GOJO Industries - Europe Ltd
Description: GOJO® Natural scrub hand cleaner contains plant-based scrubbing particles. The formulation has been developed for use on grease, oil, and tar.
UOM: 1 * 4 items
Supplier: GOJO Industries - Europe Ltd
Description: PURELL® VF+ Hygienic Hand Rub offers unprecedented sanitising while preserving healthy skin. Product benefits include a short contact time and an exce...
Catalog Number: (GOJO5796-04-EEU00)
Supplier: GOJO Industries - Europe Ltd
Description: Hygienic hand sanitising foam feels great to use and is clinically proven to maintain skin health.
UOM: 1 * 4 items
Catalog Number: (115-3475)
Supplier: GOJO Industries - Europe Ltd
Description: Spa-inspired foam hand soap is unfragranced and colourless.
UOM: 1 * 4 items
Catalog Number: (GOJO5703-12-EEU)
Supplier: GOJO Industries - Europe Ltd
Description: Three in one unscented and uncolored lotion for washing hands, body and hair.
UOM: 1 * 12 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 124
no targeter for Bottom