Finger cots, latex
Supplier: B. Braun
112-0048EA 3.9 EUR
BRAU6091032
Finger cots, latex
Suitable for protection of individual fingers during dexterous work.
  • Powder-free, smooth finish
  • Ambidextrous
  • Beaded rim
  • Very flexible
Thickness: 0,10 mm
Colour: Natural
Certifications: EN 455, AQL 1,5