You Searched For: Separating Funnels
37  results were found
SearchResultCount:"37"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass with NS-PE stopper.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, with NS-PE stopper.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, with NS-PE stopper.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, with NS-PE stopper.
Catalog Number: (LENZ400308)
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Separating Funnels, PA, Capacity: 25 ml, Socket size: 19/16 NS
UOM: 1 * 1 items
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, with NS-PE stopper.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, with NS-PE stopper.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Separating funnels, conical, made of borosilicate glass 3.3, amber coloured.
Supplier: DWK Life Sciences
Description: Made of DURAN®, borosilicate glass 3.3, clear.
Supplier: DWK Life Sciences
Description: DURAN® borosilicate glass 3.3, clear, stopcock with glass key and plastic stopper.
Supplier: Thermo Fisher Scientific
Description: PPCO translucent, TFE stopcock, PP screw closure.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear, with NS-PE stopper.
Supplier: Thermo Fisher Scientific
Description: FEP translucent, TFE stopcock, ETFE screw closure.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Separating funnels, conical, made of DURAN® borosilicate glass 3.3, with solid glass or interchangeable PTFE key and plastic stopper.
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Borosilicate 3.3 glass, delivered with NS-PE stopper.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 37
no targeter for Bottom