You Searched For: Safety Funnels
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: S.C.A.T.
Description: Collect your laboratory waste with our 'Jan' Universal Waste Hub, the smart all-in-one solution for liquid waste. It is a clever combination of lid fu...
Supplier: S.C.A.T.
Description: The strong and electrically conductive safety funnel 'ARNOLD' V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liq...
Supplier: S.C.A.T.
Description: Safety funnels with hinged lid cap, sieve and safety lance.
Supplier: Bohlender
Description: Funnel made of PE or static dissipative PE-EX for liquid waste disposal. The funnel can be safely mounted on the waste container with its freely turna...
Supplier: Bohlender
Description: Safety funnel, Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes, PE, Thread: S90
Supplier: ROETZMEIER SICHERH. BEHAELTER
Description: Stainless steel.
Supplier: S.C.A.T.
Description: The strong safety funnel 'ARNOLD' V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combina...
Supplier: Bohlender
Description: b.safe funnels with level control are made of electrically conductive dissipative PE-EX and suitable for use with flammable liquids. Electrically cond...
Supplier: Bohlender
Description: Funnel made of PE for liquid waste disposal. The funnel can be safely mounted on the waste container with its threaded connection that is safely fixed...
Supplier: NEW PIG
Description: Burpless design vents during pouring for fast, smooth flow with no dangerous splashback.
Supplier: JUSTRITE MANUFACTURING CO
Description: Galvanised steel, 18-gauge, with electrostatically applied epoxy powder coat paint.
Catalog Number: (DUEH3.80.001)
Supplier: DÃœPERTHAL GMBH & CO
Description: Stainless steel 1.4571, unpolished.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: HDPE funnels and covers, with PP closure and funnel adapter, PTFE vent membrane.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom