VWR®, Flasks, Flat Bottom, Narrow or Wide Neck
Supplier: VWR
214-1189EA 12.6 EUR
214-1189 214-1188 214-1190 214-1192 214-1191 214-1194 214-1193 214-1196 214-1195 214-1187
VWR®, Flasks, Flat Bottom, Narrow or Wide Neck
Flasks Flat Bottom Flasks
Borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area.
Certifications: ISO 1773 (narrow neck)
DIN 12347 (wide neck)