You Searched For: Flat Bottom Flasks
18  results were found
SearchResultCount:"18"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, clear.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, clear.
Sale
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass 3,3, clear.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear.
Supplier: SP Industries
Description: These glass replacement flasks are used for lyophilising equipment.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3,3, clear.
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Made of borosilicate glass.
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Made of borosilicate glass.
Catalog Number: (458-0048)
Supplier: Hach
Description: Specially designed glass flask for use with Digesdahl® digestion apparatus. Each flask is calibrated individually with a 100 ml (to contain) mark.
UOM: 1 * 1 items
Catalog Number: (WITG5.514.010)
Supplier: WITEG LABORTECHNIK GMBH
Description: DURAN® borosilicate glass 3.3 clear.
UOM: 1 * 1 items
Supplier: SciLabware
Description: PYREX®, borosilicate glass, clear, with medium necks.
Supplier: SciLabware
Description: PYREX®, borosilicate glass, clear, with short neck.
Supplier: GLASWARENFABRIK KARL HECHT
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: SciLabware
Description: PYREX®, borosilicate glass, clear.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 18
no targeter for Bottom