Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
First Aid Cabinets
https://ru.vwr.com/stibo/smallweb/std.lang.all/52/37/18945237.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (118-0227)
Supplier:  PLUM HUDSIKKERHED A/S
Description:   All in one solution for virtually any workspace. Optimal care for eye injuries and cuts.
Contents:
UOM:  1 * 1 items
Supplier:  PLUM HUDSIKKERHED A/S
Description:   QuickSafe is a flexible first aid solution that you can easily tailor to your particular needs. The cabinet takes up little more than an A3 sheet of p...
Supplier:  ORKLA CARE AB
Description:   Mini first aid cabinet with panel, 260×170×440 mm
Catalog Number: (118-0278)
Supplier:  ORKLA CARE AB
Description:   This first aid station, made of polystyrene and ABS-plastic, is especially suitable for workplaces where there is a risk of burns.
UOM:  1 * 1 items
Catalog Number: (118-0276)
Supplier:  ORKLA CARE AB
Description:   This first aid station has a transparent door with rubber edges, which protects against dust and dirt, thus making the equipment more hygienic and alw...
UOM:  1 * 1 items
Catalog Number: (118-0264)
Supplier:  ORKLA CARE AB
Description:   This first aid station has a transparent door with rubber edges, which protects against dust and dirt, thus making the equipment more hygienic and alw...
UOM:  1 * 1 items
Catalog Number: (CEDE291470)
Supplier:  ORKLA CARE AB
Description:   This small robust metal cabinet is ideal for workplaces with dirty environments, dust, moisture, etc.
UOM:  1 * 1 items
Supplier:  SOEHNGEN W. GMBH
Description:   Manufactured from high quality sheet metal 0,75 mm thick.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 9
  1  2  Next