You Searched For: First Aid Supplies
143  results were found
SearchResultCount:"143"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Made from a composite material including two plies of glass fibre matting and one layer of flame-retardant foil. For extinguishing and smothering rela...
Catalog Number: (139-0806)
Supplier: VWR Collection
Description: Laboratory safety showers should be tested once a month to check the functionality and the required water flow rate of 60 L/min. for emergency instanc...
UOM: 1 * 1 items
Supplier: Plum Safety
Description: Sterile 0,9 % Sodium chloride solution is ideal for injuries with foreign bodies like dust, dirt ect. Sterile 4,9 % Phosphate buffer solution is perfe...
Supplier: Plum Safety
Description: Sterile phosphate buffer solution (4,9%) for fast and effective neutralisation of acids and alkalis.
Catalog Number: (118-0477)
Supplier: Plum Safety
Description: This wall-mountable dispenser contains 45 elastic plasters and 5 ampoules with 20 ml of 0,9% sterile sodium chloride eyewash solution. The elastic pla...
UOM: 1 * 1 items
Catalog Number: (118-0379)
Supplier: Plum Safety
Description: This wall-mountable station is made of impact resistant plastic and equipped with a transparent cover. The station is specifically designed for use in...
UOM: 1 * 1 items
Supplier: Plum Safety
Description: QuickSafe is a flexible first aid solution that you can easily tailor to your particular needs. The cabinet takes up little more than an A3 sheet of p...
Supplier: Plum Safety
Description: Wall-mounted eyewash stations, made in hard plastic, for simple and quick first-aid in case of an eye injury.
Supplier: Plum Safety
Description: This wall station is equipped with two 1000 ml eyewash bottles, wall brackets, pictogram and mirror. The single bottle contains sterile sodium chlorid...
Supplier: Plum Safety
Description: This plaster dispenser allows quick and hygienic first-aid access, at the centre of the workplace. Pull to retrieve a ready-for-use plaster, and get b...
Supplier: Plum Safety
Description: This plaster dispenser allows quick and hygienic first-aid access, at the centre of the workplace. Pull to retrieve a ready-for-use plaster, and get b...
Catalog Number: (BRAU9080120N)
Supplier: B. Braun
Description: Silk plasters for the fixation of all types of wound dressing, e.g. Askina® Pad, Askina® gauze compresses. Also suitable for the attachment of cathete...
UOM: 1 * 12 items
Catalog Number: (BRAU9048120)
Supplier: B. Braun
Description: Elastic, wide-mesh fixation bandage made from 70% viscose and 30% polyamide. For all types of fixation dressings and light support bandages.
UOM: 1 * 20 items
Supplier: B. Braun
Description: Elastic fixation bandage made from 54% polyamide and 46% viscose for securing wound dressings and splints and for light compression bandages.
Supplier: Plum Safety
Description: Available as either an open station or closed and dust-proof wall box containing 2×500 ml plum eyewash bottles.
Supplier: B. Braun
Description: Gauze compresses with cut edges for all types of classic wound care, as an ointment or medication carrier.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 143
no targeter for Bottom