Filtration system, AcroPrep™ 24
Supplier: Pall Laboratory
516-7560EA 1590 EUR
PALL289000159
Filtration system, AcroPrep™ 24
Eliminates the time consuming one at a time syringe filter process by simultaneously filtering up to 24 samples in seconds. Ideal for use with HPLC sample preparation.
  • Can be used with any conventional 2 ml, 12×32 mm vials
  • No cross-contamination, each 1,9 ml well is individually sealed