You Searched For: Membrane Filter Units
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: DWK Life Sciences
Description: Filtration apparatus with unit made from clear DURAN® borosilicate glass 3.3, PTFE insert and anodised aluminium clamp.
Supplier: Sartorius
Description: BioSart® 100 monitors have been specifically designed for the detection and enumeration of microorganisms in beverages, water, pharmaceuticals, cosmet...
Supplier: Whatman products (Cytiva)
Description: MBS I is an excellent system for optimal microbiological control using membranes. The overall procedure time is reduced to a minimum. The design of th...
Supplier: Whatman products (Cytiva)
Description: Pressure filtration devices with a sample loading cylinder are suitable for batch filtration of samples from 20 ml. Devices without infusion cylinders...
Supplier: DWK Life Sciences
Description: These vacuum microfiltration assemblies are manufactured from 33 expansion, low extractable borosilicate glass. The 47 mm vacuum microfiltration assem...
Catalog Number: (514-0054)
Supplier: Whatman products (Cytiva)
Description: The microbiological filtration system is used for optimal microbiological control using membranes. The design of the system, which consists of an elec...
UOM: 1 * 24 items
Catalog Number: (SCERSP953820-1047)
Supplier: DWK Life Sciences
Description: This microfiltration system manufactured from 33 expansion, low extractable borosilicate glass eliminates pouring HPLC solvents into funnels. The solv...
UOM: 1 * 1 items
Catalog Number: (SCERSP953730-0000)
Supplier: DWK Life Sciences
Description: The 25 mm vacuum microfiltration assembly is designed to handle small volumes of liquids for analysis of particulate or microbiological contamination....
UOM: 1 * 1 items
Catalog Number: (SCERSP179950-4532)
Supplier: DWK Life Sciences
Description: This microfiltration adapter assembly is designed to allow connection of a 47 or 90 mm filtration support base with a straight drip tip to a bottle or...
UOM: 1 * 1 items
Supplier: Roth Carl
Description: These flow-through filters are made of PTFE. The filter membrane can be replaced easily by unscrewing the sample funnel. Plastic tubing can be connect...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom