Filtration capsules with HT Tuffryn® membrane
Supplier: Pall Laboratory
516-7515EA 0 EUR
PALL12112 PALL12122
Filtration capsules with HT Tuffryn® membrane
Final filter for reliable bacterial retention.
  • Uniform flow, maximum throughput
  • Built-in air vent
  • Integrity testable
HT Tuffryn® polysulfone membrane, PC housing, sterile
Sterilised by gamma irradiation.