Filter funnel manifolds for MicroFunnel™ filter funnels
Supplier: Pall Laboratory
516-0079EA 0 EUR
PALL15408 PALL15413 PALL15411
Filter funnel manifolds for MicroFunnel™ filter funnels
Aluminium filter funnel manifolds only for MicroFunnel™ filtration systems. Each place has an individual port control valve.