Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (15)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Filter Holders
https://ru.vwr.com/stibo/smallweb/std.lang.all/52/30/18945230.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"33"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (511-0666)
Supplier:  VWR Collection
Description:   Glass filter holder with stainless steel screen, 47 mm.
UOM:  1 * 1 items
Supplier:  Pall Laboratory
Description:   Filter holder, in-line, Stainless steel 316/303, For Ø: 25 mm
Catalog Number: (514-4001)
Supplier:  Pall Laboratory
Description:   Simplifies clean-up and degassing of mobile phase solvents and other solutions. The versatile design fits most HPLC bottles, flasks, and containers, a...
UOM:  1 * 1 items
Supplier:  Pall Laboratory
Description:   Eliminates the time consuming one at a time syringe filter process by simultaneously filtering up to 24 samples in seconds. Ideal for use with HP...
Catalog Number: (516-7607)
Supplier:  Pall Laboratory
Description:   Pressure filtration funnel, Stainless steel, 200 ml, For Ø: 47 mm, Height: 164 mm
UOM:  1 * 1 items
Supplier:  Pall Laboratory
Description:   Disc filter holders recommended for filtration of products such as aggressive chemicals, laboratory solvents, cell culture media, ophthalmics, pharmac...
Catalog Number: (516-8942)
Supplier:  Pall Laboratory
Description:   Filter holder designed for particulate filtration of small volumes (10 to 100 ml) using a syringe as final filter in gas filtration systems.
UOM:  1 * 6 items
Supplier:  Pall Laboratory
Description:   Acetal copolymer or stainless steel, PTFE seal. Reusable holder for clarification of small volumes (1 to 10 ml) using a syringe. Useful in filtering b...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 33
  1  2  3  4  5  Next