Filter funnel manifolds
Supplier: Pall Laboratory
516-9056EA 0 EUR
PALL15402 PALL15403 PALL4205
Filter funnel manifolds
Aluminium or polyurethane filter funnel manifolds. For microbiology and other applications. Each place has an individual port control valve. For 25 and 47 mm filter funnels.