You Searched For: Bottle Top Filters
34  results were found
SearchResultCount:"34"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These vacuum filter units are very useful in large volume samples separation and purification.
Supplier: VWR Collection
Description: Designed for the preparation of buffers, tissue culture media, microbiological media and other biological fluids. These vacuum filtration systems feat...
Supplier: VWR Collection
Description: These units are ideal for minimising unnecessary transfers by filtering directly into the centrifuge tube.
Supplier: VWR Collection
Description: PS funnel and collection reservoir. Systems for vacuum filtration of aqueous solutions including cell culture media, buffers or other biological fluid...
Sale
Supplier: Thermo Fisher Scientific
Description: Hydrophilic PES membrane, PS graduated housing. Ideal for biological and pharmaceutical sterilisation requirements. Universal tissue culture membrane ...
Supplier: Bohlender
Description: Vacuum filters with multi-stage hose connector with integrated lock screw for connecting vacuum tubing with Int.Ø 6 mm or 8 mm. The filters are produc...
Supplier: Thermo Fisher Scientific
Description: Surfactant-free cellulose acetate membrane (SFCA), PS funnel and collection reservoir. Ideal for cell culture, with low protein binding.
Supplier: Thermo Fisher Scientific
Description: PES membrane, PS funnel. Filters screw onto glass media bottles with 33 or 45 mm neck size.
Supplier: Thermo Fisher Scientific
Description: Triton X-free cellulose nitrate membrane, PS funnel and collection reservoir. Sterile disposable filters for cold sterilisation, clarification, prefil...
Supplier: Thermo Fisher Scientific
Description: Cellulose nitrate membrane, upper: PP, lower: PS. Ready to use for microbial analysis, water quality or environmental testing.
Catalog Number: (513-1402)
Supplier: Thermo Fisher Scientific
Description: These filters are particularly useful for large volume microbiological QC samples.
UOM: 1 * 12 items
Supplier: Thermo Fisher Scientific
Description: Nylon membrane, PS funnel and collection reservoir. For special applications. No wetting agents, the lowest in extractables, excellent alcohol resista...
Supplier: Cytiva (Formerly Pall Lab)
Description: Supor® hydrophilic polyethersulfone (PES) membrane, acrylic housing: 0,2 µm vent. Positive pressure filtration of aqueous laboratory solutions. For st...
Catalog Number: (514-0590)
Supplier: Thermo Fisher Scientific
Description: Supor® machV PES membrane, PS funnel and PP collection tube. The optimal choice for small volume tissue culture work. Offers clean, fast filtration of...
UOM: 1 * 12 items
Catalog Number: (513-1275)
Supplier: Thermo Fisher Scientific
Description: Bottle top filter, Nalgene®, PS, Membrane: PES, Sterile, Complete unit, Capacity: 115 ml, Pore size: 0,45 µm, Membrane Ø: 50 mm
UOM: 1 * 12 items
Supplier: Cytiva (Formerly Pall Lab)
Description: Supor® hydrophilic polyethersulphone (PES) membrane; PP housing. VacuCap® filtration device eliminates the need to constantly refill funnel in traditi...
Waste Reducing
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 34
no targeter for Bottom