Glass Extraction Thimble, Grade 603G, Whatman™
Supplier: Whatman products (Cytiva)
Whatman™
515-9017EA 236 EUR
515-9017 515-9005 515-9016 515-9007 533-0012 WHAT2811-308 515-9013 515-9036 515-9025 515-9031 515-9030 537-0027 515-9011
Glass Extraction Thimble, Grade 603G, Whatman™
Extractors Extraction Thimbles
Cytiva's Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems.
  • Resistant to temperatures up to 500 °C (932 °F)
  • Manufactured with walls as thin as 1 to 1,5 mm
  • Analysis of hot and acidic gases, including emission control and smoke stack gas monitoring
  • Soxhlet extraction
  • Analysis of pesticide residues
  • Determination of oil/fat content of foods
  • Analysis of oil and grease in solid wastes
Manufactured from 100% pure borosilicate glass with inorganic binder.
  • For temperatures up to 500 °C
Whatman standard glass extraction thimbles are available in a range of dimensions and wall thicknesses to suit your application.