You Searched For: Electrical Sockets
11  results were found
SearchResultCount:"11"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (115-3043)
Supplier: DESCO
Description: These plugs to fit UK European CEE (7) VII Schuko sockets as used in Belgium, France, Germany, Holland, Spain. The Europe Earth Bonding Point (EBP) pl...
UOM: 1 * 1 items
Supplier: HUBER
Description: Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue ...
Catalog Number: (SE6067)
Supplier: Cytiva
Description: Electrical socket, Banana plug
UOM: 1 * 1 items
Supplier: Agilent
Description: Electrical socket, QuickSwap restrictor set includes (2×I.Ø 0,092/0,100/0,110/0,120 mm)
Supplier: BACHMANN GMBH, ELEKTRO
Description: As for Professional, but also with:
Supplier: BACHMANN GMBH, ELEKTRO
Description: Schuko outlets in anodised aluminium, at 35° angle with nickel-plated earth and integrated child protection
Catalog Number: (139-1371)
Supplier: BACHMANN GMBH, ELEKTRO
Description: Three-Way socket, in shock resistant plastic.
UOM: 1 * 1 items
Supplier: BACHMANN GMBH, ELEKTRO
Description: Trailing socket with built-in, attractive and practical cable rewind.
Catalog Number: (139-1388)
Supplier: BACHMANN GMBH, ELEKTRO
Description: The six-way trailing socket has triple protection with a 16 A/3600 W connected load as well as surge protection (5 kA nominal leakage current).
UOM: 1 * 1 items
Catalog Number: (139-1395)
Supplier: BACHMANN GMBH, ELEKTRO
Description: Four-Way Schuko outlet with integrated one pole thermal cut-off switch.
UOM: 1 * 1 items
Supplier: BACHMANN GMBH, ELEKTRO
Description: Cable reel with four Schuko outlets 250 V~, without lid. Stable zinc-plated stand with black PVC cable (HO5VV-F G3 1,50 mm²).
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 11 of 11
no targeter for Bottom