Earmuffs, Mach 1™
Supplier: Honeywell Safety
111-0123EA 11.4 EUR
DALL1010421
Earmuffs, Mach 1™
These basic earmuffs, with adjustable headband and lightweight dielectric construction, provide adequate protection in short-term work situations.
  • Economical design provides adequate protection at a low cost
  • Extremely lightweight construction for all-day comfort
  • Dielectric construction suitable for all workplaces, especially electrical environments
SNR: 23 dB
Weight: 141 g
Certifications: EN 352-1