Soap dispensers, Eurospender 2000
Supplier: Bode
Eurospender
BODE974700EA 0 EUR
BODE974700 115-4018 BODE974690
Soap dispensers, Eurospender 2000
Soap Dispensers and Lotion Soap Dispensers
These flexible, user-friendly wall-mounted dispensers are designed for simple handling. Sampling shall be carried on the operation of the lever. The high-quality materials ensure high durability and low maintenance.
  • Easy pump change by front removal
  • All regular way wrong use, simple and quick bottle change
  • Dosing for liquid pump adjustable (approx. 0,5/1,0/15 ml per stroke); factory setting is 1,5 ml per stroke
Ordering information: Incl. mounting hardware installation and manual.
Delivery information: Accessories and spare parts are available on request. Please contact your local VWR customer service center for more information.