Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Hand Cleanser Dispensers
https://ru.vwr.com/stibo/smallweb/std.lang.all/18/37/22951837.jpg
SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"52"
  List View Easy View (new)
Sort by:
Catalog Number: (115-2098)
Supplier:  VWR Collection
Description:   Durable, wall-mountable soap dispenser for use with foam cleansers.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These VWR manual dispensers are durable, wall-mountable and for use with Deb Stoko® soaps and cleansers. Ideal for use in offices, medical rooms, labo...
Supplier:  KIMBERLY CLARK
Description:   These soap dispensers have been designed to promote hygiene, improve comfort and care.
Catalog Number: (KIMB11329)
Supplier:  KIMBERLY CLARK
Description:   This touchless cassette dispenser provides Kleenex® luxury foam soap or sanitiser. The skin care innovation in this electronic system represents high ...
UOM:  1 * 1 items
Supplier:  Deb-STOKO
Description:   Soap dispenser suitable for use with 500 ml or 1000 ml bottles − Can be used without contact.
UOM:  1 * 1 items
Catalog Number: (EVONSUN1LDSMD)
Supplier:  Deb-STOKO
Description:   This sun cream dispenser has been designed for use with Sun Protect 1000 products.
UOM:  1 * 1 items
Supplier:  Deb-STOKO
Description:   Practical white dispenser for heavy industrial applications, construction or vehicle maintenance.
UOM:  1 * 1 items
Supplier:  Deb-STOKO
Description:   This white dispenser is designed for applications in general industry, in food service and catering.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 52
  1  2  3  4  5  6  7  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping