Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "dispensers"
https://ru.vwr.com/stibo/smallweb/std.lang.all/95/51/18709551.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"233"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   ABS dispenser designed to store, dispense and cut Parafilm™, tape, labels and Tough-Tags™.
Supplier:  VWR Collection
Description:   This foil dispenser provides a good alternative to pre-cut foil squares.
Supplier:  VWR Collection
Description:   ABS containers for storing personal safety apparatus in the workplace.
Catalog Number: (115-2098)
Supplier:  VWR Collection
Description:   Durable, wall-mountable soap dispenser for use with foam cleansers.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These VWR manual dispensers are durable, wall-mountable and for use with Deb Stoko® soaps and cleansers. Ideal for use in offices, medical rooms, labo...
Supplier:  VWR Collection
Description:   Digitrate Pro™ combines modern design, with precise, accurate and convenient dispensing. Its head can be conveniently rotated 360° for operator safety...
Catalog Number: (611-3119)
Supplier:  VWR Collection
Description:   Glove boxes holder in durable resin to resist to harsh cleaning solutions.
UOM:  1 * 3 items
Supplier:  VWR Collection
Description:   These dispensers, made from white lacquered metal, are suitable for use with wipe rolls.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 233
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next