You Searched For: Cell Culture Dishes
40  results were found
SearchResultCount:"40"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Confocal dishes are suitable for fluorescent, confocal and phase-contrast microscopy experiments. These dishes combine the convenience of standard 35 ...
Sale
Supplier: VWR Collection
Description: VWR® cell and tissue culture dishes are available with a choice of surface treatments.
Supplier: VWR Collection
Description: VWR® 3-D scaffold cell culture dishes, made of polystyrene (GPPS) with polystyrene polymer scaffold, are able to simulate the three-dimensional struct...
Supplier: VWR Collection
Description: VWR® cell slides are made from medical grade, high transparency PET, which has high strength and is unbreakable. Only the surface of the cell slide ha...
Sale
Supplier: Corning
Description: PS, coated with poly-lysine.
Supplier: Corning
Description: PS coated with rat tail collagen type I, sterile, non-pyrogenic.
Catalog Number: (GREI688160_240)
Supplier: Greiner Bio-One
Description: PS, cell culture-treated, sterile.
UOM: 1 * 240 items
Supplier: Greiner Bio-One
Description: PS with integrated glass bottom, sterile. CELLviewâ„¢ glass bottom dishes combine the convenience of a standard 35 mm disposable plastic cell culture di...
Supplier: Greiner Bio-One
Description: PS, coated with rat tail collagen type I. Collagen type I is an extracellular matrix protein, which in vivo influences adhesion, migration and ...
Supplier: Corning
Description: PS. Corning® PureCoat™ surfaces are a novel family of chemically-defined, animal-free cell culture surfaces designed to enhance cell performance. A pr...
Supplier: Greiner Bio-One
Description: PS, polymer-coated. For the propagation of fastidious cells like primary or sensitive cells as well as cells cultivated under restricted growth condit...
Supplier: Greiner Bio-One
Description: These sterile, PS cell culture dishes have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modifi...
Supplier: Greiner Bio-One
Description: PS, cell culture-treated, sterile. Available with a variety of different dimensions and growth areas. The 57 cm² and 143 cm² dishes have an extra high...
Catalog Number: (734-0201)
Supplier: Corning
Description: Culture dish, Gridded, Corning® BioCoat®, PS, Ø: 150 mm, Non sterile
UOM: 1 * 5 items
Supplier: Corning
Description: PS coated with a uniform application of human fibronectin. Promotes cell attachment, spreading, proliferation and diffentiation of many cell types, pa...
Supplier: Greiner Bio-One
Description: PS, coated with poly-lysine. Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic molecules that are used to improve the adhesion of different ce...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 40
no targeter for Bottom