You Searched For: Desiccator Cabinets
43  results were found
SearchResultCount:"43"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (BOHLV1830-07)
Supplier: Bohlender
Description: Aluminium frame with 2-pane insulation (inside glass, outside acrylic glass), including four shelves made of PC, usable with up to max. 26 shelves. In...
UOM: 1 * 1 items
Supplier: Bohlender
Description: Clear polycarbonate. Each chamber has a single door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature r...
Supplier: Bohlender
Description: These transparent PMMA or borosilicate 3.3 glass cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Designed...
Supplier: Bohlender
Description: Desiccator cabinets, transparent acrylic, aluminium frame with panels made of acrylic glass, which are suitable for long-term storage.
Supplier: Palbam
Description: Bench top Nâ‚‚ purge desiccator cabinets made from stainless steel 304, units have an electropolished finish and static dissipative PVC window. Cabinets...
Supplier: Bohlender
Description: These transparent acrylic cabinets have solid 25 mm thick walls, a circular silicone seal and a vacuum gauge (0 to 760 mm of mercury).
Supplier: Bohlender
Description: Desiccator cabinets, PC ESD, have a reinforced aluminium frame with panels made of antistatic polycarbonate and an easy to read electronic hygrometer....
Supplier: Bohlender
Description: Clear, PMMA, reinforced aluminium frame with panels made of acrylic glass.
Supplier: Bohlender
Description: Transparent acrylic. Cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Designed for storing or drying humid...
Supplier: Bohlender
Description: Clear polycarbonate portable desiccator with convenient handle. Door with snap fit closing and integrated gasket ensuring easy opening and tight closu...
Supplier: Bohlender
Description: Clear polycarbonate lockable desiccators with single door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Tempera...
Supplier: Bohlender
Description: Orange polycarbonate panels reduce light incidence and protect samples from ultraviolet radiation. Door with snap-fit closing and integrated gasket en...
Supplier: Bohlender
Description: Transparent cabinets, have an reinforced aluminium frame with panels made of heat-resistant borosilicate glass 3.3. Suitable for storing hot materials...
Supplier: Bohlender
Description: Clear polycarbonate desiccators for gas filling with single door with snap fit closing and integrated gasket ensuring easy opening and tight closure.
Catalog Number: (467-0054)
Supplier: Bohlender
Description: This transparent acrylic cabinet has a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Suitable for long-term storage, fr...
UOM: 1 * 1 items
Supplier: Bohlender
Description: Opaque black polycarbonate panels reduce light incidence and protect samples from ultraviolet radiation. Door with snap fit closing and integrated gas...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 43
no targeter for Bottom