Desiccant cartridges, SP Bel-Art
Supplier: Bel-Art Products
467-4192EA 149 EUR
BELAF420450200 BELAF420450250
Desiccant cartridges, SP Bel-Art
Reusable desiccant in a rugged polyester/felt bag with Mylar® cover is ideal for use in most desiccators.
  • Gel cartridges can be regenerated in a vented oven at 150 °C for about 3 hours until blue colour reappears
  • Natrasorb® TI indicating silica gel beads are blue colour when recharged and pink when saturated
  • Can be regenerated repeatedly without losing drying performance