You Searched For: Desiccators
102  results were found
SearchResultCount:"102"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Sale
Supplier: VWR Collection
Description: Borosilicate glass 3.3
Supplier: VWR Collection
Description: Porcelain desiccator plates. Central hole Ø 20 mm, filter holes Ø approximately 5 mm. Suitable for use with any desiccators with corresponding diamete...
Sale
Supplier: DWK Life Sciences
Description: DURAN®, clear borosilicate glass 3.3. Ideal for drying and drying processes, storage under vacuum and protection from the effects of air and moisture....
Supplier: DWK Life Sciences
Description: Stainless steel 1.4301, Type 304, rust-free.
Supplier: Bohlender
Description: Desiccator cabinets, transparent acrylic, aluminium frame with panels made of acrylic glass, which are suitable for long-term storage.
Supplier: DWK Life Sciences
Description: DURAN® clear borosilicate glass 3.3, vacuum desiccator with Mobilex screw thread in the lid with stopcock. With desiccator base with flat flange, no o...
Supplier: Palbam
Description: Bench top Nâ‚‚ purge desiccator cabinets made from stainless steel 304, units have an electropolished finish and static dissipative PVC window. Cabinets...
Supplier: DWK Life Sciences
Description: DURAN® clear borosilicate glass 3.3, lid with threaded outlet, type MOBILEX (GL 32), stopcock with PTFE spindle.
Supplier: Bohlender
Description: Clear polycarbonate portable desiccator with convenient handle. Door with snap fit closing and integrated gasket ensuring easy opening and tight closu...
Catalog Number: (467-0081)
Supplier: Bohlender
Description: This transparent PMMA cabinet has a reinforced aluminium frame, forward opening door with foam rubber gasket and magnetic strip. Ideal for wide produc...
UOM: 1 * 1 items
Supplier: Bohlender
Description: These transparent PMMA or borosilicate 3.3 glass cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Designed...
Catalog Number: (BOHLV1927-07)
Supplier: Bohlender
Description: Orange acrylic cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip, orange acrylic panels reduce light inciden...
UOM: 1 * 1 items
Catalog Number: (BOHLV1938-07)
Supplier: Bohlender
Description: Aluminium frame with panels made of acrylic glass, including four shelves made of polycarbonate, tray, hygrometer and desiccant, usable with up to 38 ...
UOM: 1 * 1 items
Catalog Number: (BOHLV1894-07)
Supplier: Bohlender
Description: The Auto Big Star desiccator cabinet has an aluminium frame with panels made of acrylic glass, including two stainless steel shelves, hygrometer and m...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 102
no targeter for Bottom