You Searched For: Spectrophotometer Cuvettes
92  results were found
SearchResultCount:"92"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: High quality cuvettes manufactured from quartz or optical glass for accurate spectrophotometric analysis.
Sale
Supplier: VWR Collection
Description: Standard rectangular macro cuvettes made from Spectrosil quartz for accurate readings.
Supplier: VWR Collection
Description: Spectrophotometer cuvette, VWR®, UV-1600PC, V-3000PC UV-3100PC, UV-6300PC, Quartz, Path length: 10 mm, Height: 15 mm
Sale
Supplier: VWR Collection
Description: Disposable cuvettes in either optical polystyrene (PS) or polymethyl methacrylate (PMMA), for use with most polar solvents, acids and alkaline solutio...
Supplier: VWR Collection
Description: Square cuvette, VWR®, V-1200/UV-1600PC/UV-3100PC, Glass, Path length: 30 mm
Sale
Supplier: VWR Collection
Description: Spectrophotometer cuvette, VWR®, UV-1600PC/UV-3100PC, Quartz, Path length: 10 mm, Height: 15 mm, Volume: 500 µl
Catalog Number: (634-1067)
Supplier: VWR Collection
Description: Disposable macro cuvettes, four clear sides.
UOM: 1 * 100 items
Supplier: HELLMA
Description: Macro cuvettes for high precision analytical techniques.
Supplier: Brand
Description: Disposable macro or semi-micro cuvettes made of PS or PMMA for kinetic measurements.
Supplier: HELLMA
Description: Micro cuvettes with PTFE lids or stoppers.
Supplier: Brand
Description: Disposable cuvettes often replace expensive and sensitive cuvettes made from glass and quartz.
Supplier: HELLMA
Description: Ultra-micro cuvettes with PE stoppers.
Supplier: HELLMA
Description: Optical glass cells for special applications.
Supplier: HELLMA
Description: The cuvettes are designed with a chamber to minimise bubble formation.
Supplier: HELLMA
Description: Ultra macro cuvettes with PE stoppers designed to eliminate capillary action, which reduces filling volumes required.
Supplier: HELLMA
Description: These cuvettes with PTFE stoppers are specially designed for light-scattering measurements in the visible range of the spectrum.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 92
no targeter for Bottom