You Searched For: Adapters, Joints with Tube Connectors
26  results were found
SearchResultCount:"26"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear.
Supplier: Bohlender
Description: PTFE, white.
Supplier: BÜRKLE
Description: Adapter, 3/4 " thread inflowing side on fine thread 2" outflowing side, PP/PE, green/white, PP/PE
Supplier: Bohlender
Description: PTFE, white. For connecting tubes or tubing to vessels with ground joint. For inserting and fixing probes, thermometers, dip tubes or cables.
Supplier: ENTEGRIS INC
Description: PFA.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Bohlender
Description: This vacuum adapter is made of PTFE with O-ring made of FKM for secure placement of standard filter cartridges made of glass or plastic on vessels and...
Catalog Number: (BOHLN1656-45)
Supplier: Bohlender
Description: This vacuum adapter is made of PTFE with O-ring made of FKM for secure placement of standard filter cartridges made of glass or plastic on flasks with...
UOM: 1 * 1 items
Catalog Number: (BOHLD620-08)
Supplier: Bohlender
Description: PTFE, white. For bottles or reaction vessels with ground joint. For transferring liquids without contamination. For connection to a liquid source or a...
UOM: 1 * 1 items
Supplier: COWIE
Description: PTFE, white.
Supplier: VITLAB
Description: Adapter, PP, GL 45, GL 32
Catalog Number: (BURK5601-0120)
Supplier: BÜRKLE
Description: Universal thread adapter for nearly all drums and other containers with opening of 48 to 75 mm in diameter, regardless of the respective thread.
UOM: 1 * 1 items
Blocked
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 26
no targeter for Bottom