Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (111)
  • Supplier
  • Refine by Suppliers
    Sort by:
You Searched For:
Connectors, Tubing
153  results were found
https://ru.vwr.com/stibo/smallweb/std.lang.all/95/34/18709534.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"153"
  List View Easy View (new)
Sort by:
Supplier:  PAW BIOSCIENCE PRODUCTS, INC
Description:   Connectors, tubing, For tube I-Ø: 3,18 mm, Hose barb, non-valved, in-line, Class VI coupling body, Polysulfone
Supplier:  DWK Life Sciences
Description:   DURAN® GL 25 connector caps with multiple ports are suitable for a range of applications were small volumes of liquid need to be sampled or dispensed.
Supplier:  DWK Life Sciences
Description:   Tubing connection system with two or three top connectors are designed to be used with all sizes of DURAN® GL 45 laboratory and media bottles. These c...
Catalog Number: (ROBU18510)
Supplier:  ROBU GLASFILTER
Description:   Made of PTFE, white.
UOM:  1 * 1 items
Supplier:  DWK Life Sciences
Description:   KECK suction connectors AS M8, screw cap, white.
Supplier:  Bohlender
Description:   Tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE.
Supplier:  Bohlender
Description:   Adaptor made of PTFE for transition from female thread UNF ¼” 28 G to male thread NPT 1/8” or NPT ¼”.
Supplier:  BÜRKLE
Description:   PP or PVDF, translucent.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 153
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next