Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Connectors
https://ru.vwr.com/stibo/smallweb/std.lang.all/95/35/18709535.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"194"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   For precise connections of capillary colums in GC with inner diameter of 0,2 to 0,53 mm and outer diameter of 0,3 to 0,75 mm
Supplier:  Advanced Chromatography Technologies
Description:   PEEK HPLC column inlet / outlet connectors. These column connectors are designed for use at both the column inlet and outlet ends. They are suitable f...
Catalog Number: (C0002)
Supplier:  Advanced Chromatography Technologies
Description:   These ACE® column couplers made from stainless steel/PEEK are suitable for connecting preparative columns to stand-alone guard cartridge holders or fo...
UOM:  1 * 1 items
Supplier:  Advanced Chromatography Technologies
Description:   ACE® PEEK one-piece column couplers are ideal for connecting analytical or semi-preparative columns to stand-alone guard cartridge holders. They can a...
UOM:  1 * 1 items
Supplier:  Advanced Chromatography Technologies
Description:   ACE® capillary/nano column couplers are suitable for connecting capillary and nano LC columns to stand-alone capillary/nano guard cartridges or for ma...
UOM:  1 * 1 items
Supplier:  Advanced Chromatography Technologies
Description:   PEEK 1/32" microbore HPLC column inlet / outlet connectors. These column connectors are recommended for 1/32" microbore HPLC column connections. They ...
UOM:  1 * 10 items
Supplier:  Advanced Chromatography Technologies
Description:   Stainless steel/PEEK UHPLC column inlet / outlet connectors. These column connectors are designed to connect UHPLC columns to UHPLC systems and make o...
Supplier:  SGE a Trajan company
Description:   Column connector, Reducing union, stainless steel, for 1/8" to 1/16" OD tubing
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 194
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping