VWR®, GC Capillary Connectors
Supplier: VWR
VWR®
548-0307EA 159 EUR
548-0307
VWR®, GC Capillary Connectors
Fittings and Connectors
For precise connections of capillary colums in GC with inner diameter of 0,2 to 0,53 mm and outer diameter of 0,3 to 0,75 mm.