Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (277)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Overalls
https://ru.vwr.com/stibo/smallweb/std.lang.all/96/15/18989615.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"188"
  List View Easy View (new)
Sort by:
Supplier:  HUBEI ORIENT INT'L TRADING COR
Description:   These disposable overalls are made from non woven PP. Ideal for non critical applications in industrial, clinical or food processing environments.
Catalog Number: (113-8104)
Supplier:  HUBEI ORIENT INT'L TRADING COR
Description:   These disposable overalls made from non woven PP provide protection against dust and dirt.
UOM:  1 * 50 items
Supplier:  VWR Collection
Description:   Disposable, anti-static overall made of a microporous film with a nylon scrim between film and substrate that gives the material additional strength.
Supplier:  VWR Collection
Description:   These disposable overalls, with hood, are made from non woven PE-laminated PP fabric.
Supplier:  VWR Collection
Description:   VWR Advanced SF overalls are manufactured from spunbonded polypropylene fabric coated with breathable polyethylene film.
Supplier:  VWR Collection
Description:   These cleanroom overalls are manufactured from a latex-free, multilayer spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength...
Supplier:  VWR Collection
Description:   These breathable overalls are made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work envir...
Supplier:  VWR Collection
Description:   These overalls are made from Protect Xtra fabric which is a further development of the Protect fabric consisting of 100% polyester weave with conducti...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 188
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping