Visitors' Kit
Supplier: GM Equipment
113-0675EA 179 EUR
113-0675
Visitors' Kit
Disposable Garments
This visitor kit includes one single ply white paper mask (16 g/m²), elasticated at each end, one pair of blue PE overshoes (thickness 35 µm), one white PE visitor blouse , one size (20 g/m²), and one elasticated bouffant cap made from PP (20 g/m²).
  • Elastic items include latex
One size fits all.
Certifications: Compliant with CE standard (class 1 minor hazards).
Packaging: One individual plastic pack contains all four items.
*One size fits all