You Searched For: Disposable Garments
476  results were found
SearchResultCount:"476"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: These non woven shoe covers are made of PP with a skid-resistant CPE coating on the sole.
Sale
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: These disposable shoe covers are made of soft and durable CPE.
Sale
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: These disposable laboratory coats made from PP are ideal for non critical applications.
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: These disposable laboratory coats with shirt neck made from PP and are ideal for non critical applications.
Sale
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: These disposable PP trousers are ideal for use in combination with PP lab coats.
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: These disposable overalls are made from non woven PP. Ideal for non critical applications in industrial, clinical or food processing environments.
Sale
Catalog Number: (HOIT115802)
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: Sleeve protectors, CPE, Blue, Length: 440×220×0.03 m,
UOM: 1 * 1.000 items
Supplier: VWR Collection
Description: VWR® Maximum SF lab coats are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and ...
Supplier: VWR Collection
Description: Disposable non woven visitor coats made from PP.
Sale
Supplier: VWR Collection
Description: These disposable visitor coats, with collar, are made from non woven, SPP fabric.
Sale
Supplier: VWR Collection
Description: VWR® Advanced SF overalls are manufactured from spunbonded polypropylene fabric, coated with breathable polyethylene film.
Sale
Supplier: VWR Collection
Description: These disposable overalls, with hood, are made of breathable and durable non woven SMS fabric.
Sale
Supplier: VWR Collection
Description: VWR® Advanced laminated shoe covers are manufactured from spunbonded polypropylene fabric (SPP), coated with polyethylene film.
Sale
Supplier: VWR Collection
Description: Oversleeves made of 20 or 40 µm LDPE film.
Sale
Supplier: VWR Collection
Description: VWR® Basic SPP lab coats are manufactured from a strong, lightweight and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt...
Sale
Supplier: VWR Collection
Description: These disposable caps, with hairnet and peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, an...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 476
no targeter for Bottom