You Searched For: Spray Caps
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Spray caps for narrow neck bottles, For bottles: 1000 ml, Thread: 28 mm
Catalog Number: (RIXI8-3201-28278BL)
Supplier: RIXIUS
Description: Spray gun made up of polypropylene.
UOM: 1 * 1 items
Catalog Number: (RIXI8-2500-18-165E)
Supplier: RIXIUS
Description: Finger atomizer made of polypropylene with 2 mm sealing.
UOM: 1 * 1.400 items
Supplier: BÃœRKLE
Description: PP spray attachment.
Supplier: Kautex
Description: Wash bottle closure, LDPE, For: Narrow mouth bottles, For bottles: 200 ml, Thread: 18 mm
Supplier: KARTELL
Description: LDPE.
Catalog Number: (MUEL14.289.14)
Supplier: MUELLER & KREMPEL
Description: Spray pump, white, For: vaporiser bottles 100 ml, Thread: GL 20
UOM: 1 * 20 items
Catalog Number: (MUEL14.197.01)
Supplier: MUELLER & KREMPEL
Description: The spray caps with DIN 18 ring and protective cap are suitable for use with 10 to 100 ml dropping bottles.
UOM: 1 * 20 items
Catalog Number: (BELAF116200050)
Supplier: Bel-Art Products, a Part of SP
Description: PP, durable, chemical resistant.
UOM: 1 * 2 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom